United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 40
of
60 >
EW-23122-15
Rotary evaporator with digital display, safety-coated vertical glassware, 115VAC
Click to enlarge
More Product Details
Specifications
Accessories
Replacement parts
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-23122-15
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Evaporators
>
Rotary Evaporator Systems
>
Buchi Rotary Evaporators and Accessories
>
Buchi Rotavapor R-215 Digital Rotary Evaporators
Share
|
Rotary evaporator with digital display, safety-coated vertical glassware, 115VAC
EW-23122-15
$7,150.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Renowned systems with advanced features for ease of use
Digital display for both temperature and rotation speed
Multiple glassware configurations to meet your needs
Use these evaporators with flasks from 50 mL to 4 liters. Chemical-resistant PTFE and long-life seals provide practically maintenance-free operation and use. Sparkless, brushless motor provides safe, quiet operation with constant torque from 20 to 280 rpm.
The water/oil bath has a temperature range from 20 to 180°C. Bath has been designed with energy efficiency and safety in mind. Insulating housing prevents contact with hot surfaces; high alloy stainless steel pan is easy to clean. Heating coils are located beneath the surface of the pan to minimize maintenance and simplify cleaning. Use only uncoated flasks when using as an oil bath. Bath features wireless contacts for both power supply and communications, adding to maximum safety for both the unit and user. In the event of a power failure or fault in the bath, the flask is automatically raised to prevent overheating of the product. Each unit features digital display for bath temperature.
Choose from a wide variety of glassware to meet your needs. The diagonal condenser is used for basic distillations and is ideal for where height is limited. The vertical condenser is also used for basic distillations, however takes up less bench space. The vertical assembly also features a connection for a vapor temperature sensor. The cold-trap condenser is ideal for maximizing the condensation due to the low temperatures and no additional cooling water necessary. For distillations needing reflux, choose from either vertical or cold-trap condenser with shutoff. All glassware (except for evaporating flasks) is available with Plastic + Glas coating to minimize breakage and safety concerns.
Note:
Direct exposure of Plastic + Glas glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.Use these evaporators with flasks from 500 mL to 4 liters. Chemical-resistant PTFE and long-life seals provide practically maintenance-free operation and use. Sparkless, brushless motor provides safe, quiet operation with constant torque from 20 to 280 rpm.
The vertical condenser is used for basic distillations, while utilizing less bench space than the diagonal version. The vertical assembly also features a connection for a vapor temperature sensor. All glassware (except for evaporating flask) comes with PLASTIC+GLAS
®
to minimize breakage and safety concerns.
What's Included:
quick-action jack, 1-liter glass flasks (one receiving and one evaporating), condenser, a 5-ft power cord with three-prong plug, and a water bath with digital display.
Specifications
Bath temperature
20 to 180°C
Bath Capacity
4 L
Display
Digital
Power (Hz)
60
Dimensions
21-21/32"W x 22-41/64"H x 16-11/32"D (55 x 58 x 42 cm)
Rotation speed range
20 to 280 rpm
Speed resolution
+/- 1 rpm
Evaporation flask size
50 mL to 4 L
Watts
1300
Vacuum seal
PTFE
Joint size
24/40 (evaporating)
Lift mechanism
Quick-action jack
Condenser type
Vertical Evaporators
Glass type
Plastic and Glas coated
CE Compliance
Yes
Brand
Buchi
Power (VAC)
115
Manufacturer number
23122V100
Accessories
Qty
/
Price
Availability
Rotary Evaporator Bath Cover for small heating bath
EW-23123-56
at
$145.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Buchi Rotary Evaporator Remote Controller
EW-23123-59
at
$735.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Replacement parts
Qty
/
Price
Availability
Heating bath for Buchi Rotary Evaporators, 115V
EW-23123-50
at $1,360.00 USD/EACH
Usually ships in 22 days.
Add to Cart
Vacuum seal for Buchi Rotary Evaporators
EW-23123-60
at $57.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title