United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
5 >
EW-06750-30
Rubbermaid
®
HIPAA document container, 16 gal
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-06750-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Handling and Storage
>
Document and Waste Containers
>
Confidential Document Disposal- HIPAA Compliance
Share
|
Rubbermaid HIPAA document container, 16 gal
EW-06750-30
$155.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Non-compliance with HIPAA regulations can cost!
Ships fully assembled
Document slot positioned in front of container for easy access
Five different styles
Under the HIPAA act, your organization is liable for protecting patient or employee privacy through proper disposal of sensitive and confidential records. Implement a protection plan for document collection in your facility with containers that not only help you comply with federal regulations, but do so in a way that is attractive and convenient for the office, clinic, or laboratory. A. Model MM-06750-30 has a slim design for space efficiency and under-desk placement. Model MM-06750-32 is metal and has flat top for office product such as fax machine or printer. B. Models MM-47580-00 to 04 are constructed of fire safe steel with powder coat finish. Meets federal specification A-A-182 for fire hazard and flammable prevention. Leg levelers adjust for complete stability.
Specifications
Product Type
Waste Bins
Dimensions
20 in L x 11 in W x 31 in H
Dimensions (" W)
11
Dimensions (" H)
31
Dimensions (" L)
20
Capacity (gal.)
16
Brand
Rubbermaid
Manufacturer number
9W25
Model
9W25
Technical Resources
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title