TBS SHUR/Wave SW-120 Laboratory Microwave,120VAC, 50/60Hz, 20A

Representative image only.
TBS Mfr # SW-120 – Item # EW-48578-01
Get routine specimen fixation and processing results in as little as 10 minutes!
  • Fast, reliable results in a fraction of the time
  • Maintains uncompromised morphology and antigenicity
  • No xylene or hazardous chemicals required—save on exposure, use and disposal of reagents!
  • Obtain superior sectioning characteristics
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Internal Width (in)13
  • Internal Height (in)7
  • Internal Depth (in)12
  • Internal Width (cm)33.02
  • Internal Height (cm)17.78
  • Internal Depth (cm)30.48
  • Power (VAC)120
  • Power (amps)20
  • Power (Hz)50/60
  • DescriptionLaboratory Microwave,120VAC, 50/60Hz, 20A
  • Warranty1 year
1 Year Warranty

Key Features

  • Fast, reliable results in a fraction of the time
  • Maintains uncompromised morphology and antigenicity
  • No xylene or hazardous chemicals required—save on exposure, use and disposal of reagents!
  • Obtain superior sectioning characteristics
  • 1 year warranty

More About this Item

The SHUR/Wave laboratory microwave was specifically designed to accelerate lengthy procedures involving tissue fixation and processing. Also great for specimen stabilization and hardening, epitope retrieval, and acceleration of specimen decalcification and staining. Creating, storing, and running programs can be mastered in a matter of minutes via the 8" touch-screen color membrane display and user-friendly controls. Programmable logic controller (PLC) allows at least 30 programs, each with up to 12 steps, to be named, stored, and run. The graphic display supplies readouts of time versus temperature. Easily run samples in one of five selectable languages including English, German, Spanish, French or Italian.

Microwave features numerous built-in safety devices to protect the operator and specimen, including a temperature controller and a ventilation system that can be ducted into your fume hood or exhaust system. Quality control program with password protection can document temperature, adequate line voltage and the dualmagnetron power output. The flexible thermocouple temperature probe, along with
proportional, integral, differential (PID) controller monitors the temperature to assure that the production of microwaves is proportioned to maintain temperature within user guidelines — eliminates the need for water coolers or hazardous chemicals, ensuring specimen safety. Order histoprocessing racks and containers separately below. Additional accessories and replacement components available at www.coleparmer.com.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat