United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
5 >
EW-86292-04
Kit 86292-08 shown
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86292-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Spill Control
>
Spill Cleanup
>
Spill-X Spill Control Products
>
Spill Treatment Kits
Share
|
SPILL-X-S Solvent Spill Treatment Kit
EW-86292-04
$487.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Treat chemical spills in the lab or on the plant floor with a single step!
What's included in each kit:
Spill Kit Treatment Guide with instructions for start-to-finish cleanup
Six easy-to-handle polypropylene containers for pouring or shaker-type application of agent
Convenient green plastic case for compact storage
One pair of rubber gloves and safety goggles
100 pH test papers with color guide
Scraper, two pick-up pans, and six disposal bags
Easy-to-use shaker containers with instructions make cleanup quick. Order agent refills separately.
Spill-X-S Solvent Adsorbent
reduces vapors released during cleanup and elevates flash point of flammable organic spills. Treats spills of many common solvents, reagents, and fuels. Does not treat organic peroxides or hydrazine compounds.
Specifications
Description
SPILL-X-S®
Agent color
Black
Chemical treated
Solvent
Brand
SPILL-X-S
Manufacturer number
78778
Model
78778
Accessories
Qty
/
Price
Availability
SPILL-X-S Solvent Agent Refill, 6 containers
EW-86292-54
at
$379.00 USD/PACKAGE
Usually ships in 7 days.
Add to Cart
SPILL-X-S Bulk Agent
EW-86293-54
at
$537.00 USD/EACH
Usually ships in 11 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Isolating Genomic DNA from Whole Blood
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title