United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
Sanitary Pressure Gauge 0 60 PSI 3 4
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-68056-74
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17040-10
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 1% or >1%)
$99.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Sanitary Gauges, Transducers, and In-Line Seals
>
Wika 3A Sanitary Gauge
Share
|
Sanitary Pressure Gauge, 0-60 PSI, 3/4
EW-68056-74
$492.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Manufactured for clean–in–place (CIP) applications
Glycerin-fill reduces fluctuations for easier reading of gauge
Autoclavable gauges are available - call our Application Specialists @ 800-323-4340 x 3
Gauges all-welded assembly contains an external flush diaphragm and is ideal for applications in the food, beverage, pharmaceutical, and biotechnology industries. Gauges are constructed of 316 stainless steel and have a 3/4" Tri-Clamp
®
process connection welded to the gauge. Assembly meets the criteria set forth by 3A standards. Gauge case and ring are 304 stainless steel and has a polycarbonate window.
Specifications
Product Type
Sanitary pressure gauges
Media compatibility
any media compatible with 316L SS
Wetted materials
316 SS
Accuracy
±3-2-3% of span
Unit of Measurement
psi
Resolution
1 psi
Range
0 to 60 psi
Process connection
3/4" Tri-clamp, lower
Liquid fill
Glycerin
Dial size (" Dia)
2.5
Case
304 SS
Lens material
Polycarbonate
Display
analog dial
Brand
Wika
Manufacturer number
4324005
Model
M932.25
Accessories
Qty
/
Price
Availability
Gauge Guard Isolator Sanitary Gasket, Platinum-cured Silicone, 3/4" Clamp Size
EW-30565-02
at
$43.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Gauge Guard Protector Cap, Platinum-cured Silicone, 3/4" Clamp Size
EW-30565-12
at
$70.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Gauge Guard Isolator Sanitary Gasket, Fkm, 3/4" Clamp Size
EW-30565-22
at
$41.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Gauge Guard Isolator Sanitary Gasket, Epdm, 3/4" Clamp Size
EW-30565-62
at
$38.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 1% or >1%)
EW-17040-10
at $99.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Sanitary and Sterile Products Brochure (359 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
The Sanitary Instrumentation Primer
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Show All...
Pressure Unit Conversion Constants
The Hidden Costs of Mercury-Filled Vacuum Gauges
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title