Sartorius MA Moisture Determination Balance, Ceramic Infrared heater, 35g x 0.001%, 115VAC
YO-11213-98
$3,250.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Feature automatic, semi-automatic, and timed drying modes
  • User-friendly programming operations with onscreen icons
  • Space-saving design conserves valuable bench top space
  • Unit has a metal IR heating element and a user-friendly, rugged design
  • Concise display shows the user important information at a single glance
  • Easy-to-understand icons guide you in three steps from taring the sample pan to starting the measurement
  • Internal memory for one moisture analysis program
The MA35 has a rugged design and is user-friendly. The concise display shows the user all important information at a single glance. Easy-to-understand icons guide you in three steps from taring the sample pan to starting the measurement. To make the MA35 even more user-friendly, we have done away with seldom-used programming options without compromising flexibility or measurement accuracy. Samples are heated by two powerful tubular-shaped metal heating elements (also called dark radiators), and an integrated reflector for uniform heat distribution.
What's Included:
80 aluminum weighing pans and a 5-ft (1.5 m) power cord
Product Type Moisture Balance
Calibration External
Capacity (g) 35
Capacity (oz) 1.2
Heater type Infrared
Housing dimensions 8-3/8" x 7-1/8" x 12-3/4" (21.3 x 18.1 x 32.4 cm)
Pan size (dia) 3-1/2" (8.9 cm)
Power (VAC) 115
Readability 0.001%
Repeatability 0.2% (=1 g), 0.05% (=5 g)
Temperature range (º C) (low) 40
Temperature range (º C) (high) 160
Timer (min) 0.1 to 99.99
Weighing units % moisture, % solid, % moisture dry, mg weight loss, or grams of residue
Output RS-232
Brand Sartorius
Accessories
Qty/Price
Availability
at $82.00 USD/EACH
Usually ships in 12 days.
at $425.00 USD/EACH
Usually ships in 6 days.
Related products
Qty/Price
Availability
at $45.00 USD/EACH
In stock.
at $45.00 USD/BOX of 50
In stock.
Calibrations
Qty/Price
Availability
at $140.00 USD/EACH
Usually ships in 8 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Do you have questions about this product?
get answers from real customers and in-house experts with AnswerBox.
2 Questions | 2 Answers
Displaying questions 1-2
  • Q:
    how do you calibrate this machine. Is there any reference standards to compare results?
    Asked on 11/28/2011 by eve from anaheim, ca
    1 answer
    • Staff Reviewer
      A:
      Calibration is much like a regular balance with an external mass. The external calibration mass is not included, this unit accepts a 30 g mass, so something like part number 11958-10 would work.
      Answered on 11/30/2011 by Jen from Technical Support
  • Q:
    will this work in a standard outlet?
    Asked on 6/16/2011 by Goonie from Waukegan
    1 answer
    • Staff Reviewer
      A:
      Yes, this is a 115V unit with standard domestic US plug.
      Answered on 6/16/2011 by Jen from Technical Support
Displaying questions 1-2
Do you have a question about this product? 
Product Reviews
REVIEW SNAPSHOT®
by PowerReviews
Sartorius<reg> Moisture Analyzer, 115 VAC, Basic, Ceramic IR Radiator, 35g
4.5
(based on 2 reviews)
Reviewed by 2 customers
Sort by
Displaying reviews 1-2
Back to top
5.0
Reliable
By OZZMAN1163
from New Tazewell, TN
Verified Reviewer
Pros
  • Accurate
  • Durable
  • Easy To Use
  • Well Calibrated
  • Works Properly
Cons
    Best Uses
    • In The Field
    Comments about Sartorius<reg> Moisture Analyzer, 115 VAC, Basic, Ceramic IR Radiator, 35g:
    This is the second unit we purchased. The first unit finally died after many years of service. Very nice unit.
    4.0
    Nice unit for the money.
    By Charlie
    from Michigan
    About Me Lab Assistant
    Pros
    • Accurate
    • Easy To Use
    • Well Calibrated
    • Works Properly
    Cons
    • No Level Bubble
    • No Multiple Setups
    Best Uses
    • In The Lab
    • Research
    Comments about Sartorius<reg> Moisture Analyzer, 115 VAC, Basic, Ceramic IR Radiator, 35g:
    This is an updated version of an existing unit we use. The older unit is easy to switch test types. Also, this doesn't have a leveling bubble like the older version.
    Displaying reviews 1-2
    Back to top
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title