Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Schott 500 Electronic Piston Burette

EW-24906-02
$4,790.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Ideal for manual titrations, accurate dosing of small and large volumes and the preparation of solutions
  • Highly visable, full color display can be viewed from a distance
  • Reagent data stored in intelligent exchangeable modules
  • Connection flexibility for printer and/or an analytical balances
  • Complete remote control via RS232 or USB interface
The motor-driven piston burette has intelligent exchangable dosing modules to ensure high precision and method flexibility. System includes preprogrammed standard methods and fifteen user-defined methods. Maximum dosing speed is 100 mL/min.

The intelligent exchangeable modules for with 5, 10, 20, and 50 mL volume store reagent and unit data on an integrated RFID chip for easy changes of titrants/solutions. Burette size (mL), titrant name, titrant concentration, expiration date and more are automatically uploaded with the change of modules.

Flexible configuration with three USB and two RS232 ports for connections with magnetic stirrer, manual controller, printer, keyboard, data storage device, balance, computer, and USB expansion hub for up to 16 devices. Automatic weight recording when balance is connected.
What's Included:
20-mL glass cylinder exchange module, manual mouse controller, magnetic stirrer, titration stand rod, titration clamp, and 100 to 240 VAC power supply.
Dispensing accuracy ±0.1 to 0.3% total volume
Capacity (mL) 20mL; 5 to 50 with additional modules
Repeatability ±0.15% of span
Display LCD, 3 1/2" H
Display resolution 0.001
Accuracy ±0.15% of span
Power (VAC) 100 to 240 VAC
Output USB abd RS-232
Power (Hz) 50/60 Hz
Brand Schott
Manufacturer number 285220220
Model 500

Accessories
Qty/Price
Availability
Dosing module, 5ml
EW-24906-54

at $1,265.00 USD/EACH

Call 1-888-358-4705 for availability.
Dosing module, 10ml
EW-24906-56

at $1,275.00 USD/EACH

Call 1-888-358-4705 for availability.
Dosing module, 20ml
EW-24906-58

at $1,275.00 USD/EACH

Call 1-888-358-4705 for availability.

at $33.00 USD/EACH

In stock.
Dosing module, 50ml
EW-24906-62

at $1,275.00 USD/EACH

Call 1-888-358-4705 for availability.
Magnetic stirrer
EW-24906-64

at $745.00 USD/EACH

Call 1-888-358-4705 for availability.




Technical Resources
  • Titronic 500 manual
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title