United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Schott automatic volumetric titrator 20 mL capacity 115 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-24906-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Titrators
>
Volumetric and Potentiometric
>
Schott Automatic Volumetric Titrator
Share
|
Schott automatic volumetric titrator, 20-mL capacity, 115 VAC
EW-24906-20
$3,950.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Automatically titrate to your endpoints using a keypad for digital control
Eliminates the difficult task of reading a burette and judging color changes at endpoints
Automatic titrators provide added convenience over manual methods
Autotitrator stops at your preselected pH endpoint or equivalence point every time. There’s no need to worry about overshooting your endpoint. Use the keypad to program pre-titration volume and speed.
Use the autotitrator with any combination pH electrode. The pH buffer solutions are stored in memory to make calibration easy. Access the eight pH buffers—2.00, 4.00, 4.01, 6.87, 7.00, 9.18, 10.00, and 12.45through the keypad.
For high-accuracy dosing, the glass cylinder features an automatic valve system. The integrated system fills without prompting for easy dosing. For incremental dosing operations, adjust the volume
and waiting time between volume increments.
Connect FEP tubing (24906-60) and bottletop adapter (24906-55) to 1-L reagent reservoir (24906-52) and titrator to add reagent. Order reservoir, adapter, and tubing separately.
What's Included:
glass combination pH electrode, magnetic stirrer, titration stand rod, titration tip holder, titration tip with tubing, glass cylinder and valve system, and 3-ft (0.9-m) cord with two-prong plug.
Specifications
Product Type
Volumetric Titrators
Dispensing accuracy
±0.15% of total volume
Repeatability
±0.15% of span
Display resolution
0.01 mL
Accuracy
±0.15% of span
Power (VAC)
115
Output
RS-232
Brand
Schott
Manufacturer number
285212831
Accessories
Qty
/
Price
Availability
Reagent reservoir, one-liter, for titrators 24906-00 to -25
EW-24906-52
at
$65.00 USD/EACH
In stock.
Add to Cart
Bottle top adapter
EW-24906-55
at
$134.00 USD/EACH
In stock.
Add to Cart
FEP tubing, 40-inch long, for titrators 24906-00 to -25
EW-24906-60
at
$33.00 USD/EACH
In stock.
Add to Cart
Replacement silver electrode
EW-24906-75
at
$410.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Glass electrodes with removable cable
EW-55503-00
at
$193.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Bioethanol Test Method Catalog
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title