United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Scienceware 36 handle 500ml dipper
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06282-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Ladles and Dippers
>
Scienceware Dippers
Share
|
Scienceware 36" handle 500ml dipper
EW-06282-00
$51.00 USD/PACKAGE of 1
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$45.90/PACKAGE of 1
30
15%
$43.35/PACKAGE of 1
120
20%
$40.80/PACKAGE of 1
In stock.
Qty
:
Add to Cart
Save to shopping list
Lightweight, yet sturdy, dippers feature a polypropylene (PP) cup that screws onto the handleorder extra cups for your multiple sampling applications. Two pouring spouts on the cup allow you to pour from either side. Long handles make these dippers ideal for sampling from large tanks, vats, streams, and lakes. High-density polyethylene (HDPE) handles are ribbed for a sure grip; end has hook for hanging.
Specifications
Product Type
Ladles & Dippers
Dimensions
3.875 in dipper diameter
Handle length
36"
Sterile
Yes
Brand
Scienceware
Capacity (mL)
500
Quantity
1
Material
HDPE handle, PP cup
Accessories
Qty
/
Price
Availability
Extra/replacement screw-on Polypropylene Cup for 06282-00 and 06282-05 Scienceware dippers
EW-06282-50
at
$13.00 USD/PACKAGE of 1
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Physical Properties of Plastics
Physical Properties of Glass
Physical Properties of Stainless Steel
Show All...
E/S Portable Sampler
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title