Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

SealPlate films, 100/pk

EW-13024-00
$53.25 USD/PACKAGE

  • Prevents evaporation and protects sample during processing, storage, and shipment
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
SealPlate 50 µm polyester films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover. Secure sealing of all wells eliminates edge effects in sensitive ELISA assays. Films can also be used with tissue culture plates for short-term storage, incubation, and containment of biohazards. Films are non-pierceable. Each film measures 79.4 x 146.0 mm.

Ministrips films (13024-04 and 06) are identical to the microplate films, but are designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates whenever rows must be selectively protected or accessed. Also used as a secondary seal to cover films that have been pierced. Each film measures 19.0 x 103.2 mm with two remo

Product Type Microplates
Brand Excel Scientific SealPlate
Manufacturer number 100-SEAL-PLT





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title