United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
SelecTone Audible Alarm 24 VAC VDC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10406-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Alarms
>
Federal Signal SelecTone Audible Alarm
Share
|
SelecTone Audible Alarm, 24 VAC/VDC
EW-10406-02
$251.73 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Emit emergency signals from one of 32 tones
Tone volumes from 64 to 88dB at 10 feet
Use these audible alarms for indoor or outdoor alarming applications. Compact, low profile design makes installation ideal in factories, plants, and offices. Solid state circuitry ensures reliable and continuous operation. Required Universal Tone Module (sold separately below) offers a choice of 32 alarm tones select desired tone via dip switch. The module fits into the alarm for easy plug-in installation. Plastic cover impregnated with stainless steel fibers shields circuitry from RFI and EMI.
Specifications
Product Type
Alarms
Dimensions (" W)
4-1/2
Dimensions (" H)
4-1/2
Dimensions (" D)
3-1/2
Power (VDC)
24
Brand
SelecTone
Model
50GC-024BG
Manufacturer number
50GC-024BG
Power (VAC)
24
Accessories
Qty
/
Price
Availability
Universal Tone Module
EW-10406-10
at
$117.85 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Alarm/Status Indicators: An Overview
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title