dummy
EW-07104-50
- No valves to clog or wear out
- Build your own pump systems using interchangeable heads, drives, and accessories
- Pump fluids with viscosities up to 500 cp
- Low dead volume
Choose stainless steel (SS) materials for added thermal and physical strength characteristics; choose ceramic materials for use with common abrasives. Sanitary high-flow models are ideal for pumping food products. Pump heads with ceramic piston and carbon cylinder liner are great for gas and liquid transfer.
To order a complete pump assembly, first select a pump head with your desired flow rate and piston diameter. Next, select a drive that meets your flow rate and power requirements.
NOTE: To connect the Stainless steel pump head module to 1/4" tubing please use adapter 07104-81. To connect to 3/8" ID tubing, please select adapter 07104-83. These are sold in quantities of one and one will normally need to have two to complete their system.
| Product Type |
Piston Pump Heads |
| Wetted parts |
316 SS diaphragm & cylinders, carbon cylinder liner |
| Max pressure |
100 psi |
| Max viscosity (cp) |
500 |
| Max flow rate (mL/min) |
48 |
| Pump type |
Piston |
| Diameter (piston) |
1/8" |
| Strokes per minute (Max) |
600 |
| Port size |
1/4" NPT(F) |
| Duty cycle |
continuous |
| Suction lift (wet) |
15 ft |
| Max temperature (º C) |
60 |
| Max temperature (º F) |
140 |
| Accuracy |
±0.1% |
| Brand |
Masterflex |
| Manufacturer number |
Q0SSY |
at $14.25 USD/PACKAGE of 10
In stock.
at $14.25 USD/PACKAGE of 10
In stock.
at $61.00 USD/EACH
In stock.
at $395.00 USD/EACH
In stock.
at $37.00 USD/EACH
In stock.
at $190.00 USD/EACH
Usually ships in 6 days.
at $775.00 USD/EACH
Call 1-888-358-4705 for availability.
at $70.00 USD/PACKAGE of 2
Call 1-888-358-4705 for availability.
at $40.00 USD/PACKAGE of 2
In stock.
at $49.00 USD/PACKAGE of 2
Usually ships in 7 days.
at $40.00 USD/PACKAGE of 2
Call 1-888-358-4705 for availability.
at $37.00 USD/PACKAGE of 2
Usually ships in 5 days.
at $60.00 USD/EACH
Usually ships in 5 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection