United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
EW-07117-66
Series R Valveless piston pump heads
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07117-66
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
>
Pumps - Piston Pumps - Valveless
>
Series R Valveless Piston Pump Heads
Share
|
Series R Valveless Piston Pump Head, 0.05 mL/stroke
EW-07117-66
$495.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Compatible with Masterflex
®
L/S
®
or series Q drive
No valves to clog or wear out
Use for low-flow applications
Kynar
®
pump body withstands harsh chemicals.
Series R pump heads come equipped with rotating pistons to prevent clogging—ideal for pumping viscous liquids to 500 cp
Compression fittings accept 1/4" OD tubing. For a complete assembly, order series R pump head and a Masterflex
®
L/S™ or series Q drives. Adapter kits let you attach pump heads to drives.
Flow characteriestics may vary with different fluids.
Specifications
Product Type
Piston Metering Pumps
Wetted parts
Kynar® compression fittings & cylinder, ceramic (alumina) piston and cylinder liner
Max pressure
100 psi
Max viscosity (cp)
500
Min flow rate (mL/min)
0
Max flow rate (mL/min)
90
Dimensions
3-1/2"L x 2-1/4" dia
Pump type
Piston
mL/stroke
0.05
Diameter (piston)
3/16"
Port size
1/4" OD compression fittings
Suction lift (wet)
15 ft
Max temperature (º C)
100
Max temperature (º F)
212
Accuracy
±1%
Accessories
Qty
/
Price
Availability
Series R pump head adapter kit for models 7117-66, -68; adapts to any Masterflex L/S drive
EW-07115-99
at
$125.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Series R pump head adapter kit for models 7117-66, -68; adapts to any Series Q drive
EW-07117-99
at
$30.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Basics of Cartridge Filtration
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
The Ins and Outs of Metering Pumps
Metering Pump Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title