United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
EW-36850-02
Seward Stomacher 80 Lab Blenders, compact, 230VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36850-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Blenders and Paddle Blenders
>
Reciprocating Paddle Blenders
>
Seward Stomacher Lab Blenders
Share
|
Seward Stomacher 80 Lab Blenders, compact, 230VAC
EW-36850-02
$5,475.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Blend and homogenize within sealed processing bagseliminating cross-contamination and cleaning between samples
Small footprint offers convenient placement on your lab bench
Increase productivityno need to clean equipment between processing, simply discard used sterile processing bags after use
Paddle design processes materials that cannot be handled by conventional lab blenders
The unique action of the Stomacher uses paddles to homogenize samples through a combination of crushing and stirring forces. Gamma-irradiated sterile disposable bags (order separately) allow you to homogenize and transport samples without changing containers or handling sterile samples.
Stomacher 80 Biomaster
is the smallest paddle blender on the market—it can fit into safety cabinets to reduce your exposure to hazardous materials. Three time settings plus continuous cycle help processes sample volumes from 5 to 80 mL. Install the optional door upgrade kit and process micro sample volumes as small as 250 mL; also allows you to adjust paddle speed, pressure, and processing timeright from the dooruseful when processing tissue and other samples without diluents or solvents.
Specifications
Product Type
Paddle Blenders
Description
Stomacher 80 Biomaster
Power (VAC)
230
Color
White and Blue
Dimensions
11.5 in W x 7.5 in H x 12.75 in D (29.2 x 19.1 x 32.4 cm)
Brand
Seward
Manufacturer number
0080/000/EU
Technical Resources
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title