United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Shore D digital hardness tester
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-59790-06
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Hardness Testers
>
Shore D Model of Digital Hardness Tester
Share
|
Shore D digital hardness tester
EW-59790-06
$1,933.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Conform to ASTM D2240, DIN 53505, ISO R/868, and JIS K7215 Shore scale hardness testing standards
These hardness testers are available in low-cost analog or digital versions. Simply position tester on sample and apply constant pressure. Choose an economical analog model or a digital model with 3 1/2-digit LCD.
Shore D models 59790-02 and -06 have a 30° sharp-point penetrator for testing hard materials such as hard rubber, acrylic, glass, and dense wood.
Each hardness tester includes reference test block, instructions, and hard carrying case.
Order bench stand 59790-50 separately to increase the accuracy and repeatability of your tests.
What's Included:
reference test block, instructions, and hard carrying case.
Specifications
Product Type
Durometer
Min sample thickness (in.)
0.12
Min sample thickness (mm)
3
Range
0 to 99 HS (Shore units)
Resolution
0.5 HS
Measured area
0.59" x 1.89" (15 mm x 48 mm
Dimensions (" W)
3.625
Dimensions (" H)
7.125
Dimensions (" D)
1.3125
Dimensions (" L)
3.625
Display
LCD
Power
two 1.5 V button cells (included)
Accessories
Qty
/
Price
Availability
Battery; Silver Oxide; 1.5 V; 150 mAh (Typ.)
EW-09376-14
at
$3.80 USD/EACH
Usually ships in 7 days or sooner.
Add to Cart
Bench stand for 59790-series hardness testers
EW-59790-50
at
$1,872.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title