United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
11 >
EW-30565-07
SILICONE GASKET ISOLATOR, 3/4"
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-30565-07
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fittings
>
Gaskets
>
Sanitary Gaskets
>
Gaskets for Sanitary Clamp Fittings
>
Gauge Guard Protector and Isolator Seals
Share
|
Silicone Gasket Isolator, 3/4"
EW-30565-07
$64.00 USD/EACH
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
Extend the life and accuracy of your expensive and fragile instruments
Manufactured to meet USP Class VI certification
Animal derived ingredient free
Gauge Guard Protectors
safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals.
Gauge Guard Isolator Seals
combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.
Specifications
Product Type
Guage Guards/Gasket Isolators
Model
40GGI-PX-075
Brand
Gauge Guard Protector
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title