United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
Sludge Judge Ultra Sampler System
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-05465-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Samplers
>
Water and Industrial Liquid Sampling
>
Sludge Sampling and Settled Solids Measurement
>
Sludge Judge Sludge Samplers
Share
|
Sludge Judge Ultra Sampler System
EW-05465-40
$165.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Measure settled solids in sewage treatment plants, chemical plants, or food processing facilities
Take accurate, noncaustic measurement of settable solids, 5% or less
A variety of Sludge Judge samplers are available to fit your sampling needs. Order complete assemblies to receive three 5-ft (1.5-m) sections that screw together, or separate component pieces if customized sampler lengths are desired. The bottom piece has a ball valve that allows liquid in when lowered into the sample and seals the sample when raised from the liquid. The middle section connects the top and bottom sections, order additional middle sections to increase the overall length. The top section has a nylon rope that aids in sampling. All sections feature 1-ft (0.3-m) increments which allow you to easily indentify how much sludge is in the sampler body.
Basic Sludge Judge and Sludge Judge Ultra versions have 3/4" (1.9 cm) diameter pipe sections and hold approximately 3 oz (89 mL) of fluid per foot. Sludge Judge II version has 1-1/4” (3.2 cm) diameter pipe sections doubling the overall capacity of the sampler. Sludge Judge Ultra additionally features an ultraviolet stabilizer which reduces material degradation from UV light.
Specifications
Brand
Judge
Manufacturer number
B01386WA
Accessories
Qty
/
Price
Availability
Sludge Judge Ultra Replacement Bottom Section with Valve
EW-05465-41
at
$76.00 USD/EACH
In stock.
Add to Cart
Sludge Judge Ultra Replacement Middle/Extension Section
EW-05465-42
at
$54.00 USD/EACH
In stock.
Add to Cart
Sludge Judge Ultra Replacement Top Section with Rope
EW-05465-43
at
$55.00 USD/EACH
Usually ships in 7 days.
Add to Cart
STORAGE BAG FOR 05465-70
EW-05465-77
at
$35.00 USD/EACH
In stock.
Add to Cart
CLEANING BRUSH F/05465-70
EW-05465-78
at
$33.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
E/S Portable Sampler
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title