Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Small Kimwipes wipe dispenser 4.625, White

Item# EW-63528-10

$18.75 USD/EACH

Are you a reseller or buying in bulk? Call us today.

  • Keep disposable supplies where they're needed
  • Free valuable drawer space for storage

Wipe Dispensers are ready to mount near instruments, workstations, hoods, and benches. Simply peel adhesive tape backing and press on.
Product Type Organizer
Color White
Qty/ea 1
No. of drawers or compartments 1
Dimensions (" W) 4.625
Dimensions (" H) 3
Dimensions (" D) 4.75

Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      can this dispenser be hung on a wall?
      Asked on 5/13/2011 by Anonymous

      1 answer

      • Staff Reviewer


        Yes, these come with a double sided adhesive and can be hung on a wall.

        Answered on 5/16/2011 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought