United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 174
of
2494 >
Representative photo only
Sod Hydrox Nfepbpfccjp 12kg
Click to enlarge
More Product Details
Specifications
Technical Resources
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-88081-32
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Chemicals, Reagents, & Lubricants
>
Specialty Chemicals
Share
|
Sod Hydrox Nfepbpfccjp 12kg
EW-88081-32
$813.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Chemical Reagents
Manufacturer number
S39212
Technical Resources
MSDS 1 (135 B)
Related technical articles (Roll over for summary)
ChemicalsSpecial Ordering Information
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title