Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Sodium Hydroxide (Granular/NF/FCC/EP/BP/JP) (2.5kg)

Fisher Chemical  |   Mfr# S392212  |   Item# EW-88024-12

$350.00 USD/EACH
$245.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.




    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Inorganic Chemicals
    Purity grade NF or FCC or EP or BP or JP
    CAS Number 1310-73-2 and 497-19-8
    Form Granular
    Formula weight 40.00
    Empirical formula NaOH
    Packaging Poly Bottle
    Synonyms Lye, Caustic Soda
    Storage symbol White
    UN number UN1823
    Hazard symbol Corrosive
    Quantity (g) 2500
    DOT information Class 8, Packing Group II
    DOT description Sodium hydroxide, solid
    Brand Fisher Chemical
    Manufacturer number S392212
    Qty/ea 1
    Class Reagent Acids and Bases





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews