Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Sodium Hypochlorite Solution (4.0-6%/Laboratory) (4 liter)

Fisher Chemical  |   Mfr# SS2904  |   Item# EW-88024-62

$205.00 USD/EACH

Discontinued
Qty

  • Assay—4-6%


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade Laboratory
CAS Number 7681-52-9
Form Liquid
Formula weight 74.44
Empirical formula NaOCl
Packaging Poly Bottle
Storage symbol White
UN number UN 1791
Hazard symbol Corrosive
Quantity (mL) 4000
DOT information DOT Class 8
DOT description Corrosive
Brand Fisher Chemical
Manufacturer number SS2904





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews