Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Sodium chlorite, unstabilized tech., 80% (500g)

Acros Organics  |   Mfr# AC223235000  |   Item# EW-88239-18

$92.00 USD/EACH

Discontinued
Qty


    Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    CAS Number 7758-19-2
    Formula weight 90.44
    Empirical formula ClNaO2
    Linear formula NaClO2
    Hazard symbol Oxidizing, Toxic
    Brand Acros Organics
    Manufacturer number AC223235000





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    3 Questions | 3 Answers
    Displaying questions 1-3
    • Q:

      if the assay is 80% what describes the remainder sodium Chloride and what else?
      Asked on 8/3/2010 by Beaker from Corvallis, Oregon

      1 answer

      • STAFF EXPERT

        A:

        This product is 80% sodium chlorite and 20% sodium chloride.

        Answered on 8/5/2010 by Jack from Technical Support
    • Q:

      Is this available in tablet form?
      Asked on 7/19/2010 by Barry O'Neill from Cambridge, MA

      1 answer

      • STAFF EXPERT

        A:

        This is not available in tablet form.

        Answered on 7/22/2010 by David from Tech Support
    • Q:

      is the sodium chlorite FCC grade
      Asked on 6/29/2009 by Anonymous

      1 answer

      • STAFF EXPERT

        A:

        This item is Technical grade. With help in finding a chemical that will fit your application, please contact our technical application specialists for additional assistance.

        Answered on 7/1/2009 by Josh from Technical Support
    Displaying questions 1-3

    Do you have a question about this product? 

    Product Reviews