Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

SoftGUARD Hand Cream, 16 oz Bottle

EW-17790-10
$122.00 USD/CASE of 6

Usually ships in 11 days.
Qty

  • Nongreasy formula absorbs immediately
Relieves irritation, prevent chapping, and protect your hands from frequent washing, solvents, and powdered gloves. Unscented
Product Type Hand Cream
Format Pump
Packaging 16 oz bottle, 6 per case
Brand SoftGUARD
Manufacturer number N9340A-ADP
Model N9340A-ADP





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title