United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
Representative photo only
Spectra Por 3 Membranes flat width 45mm diameter 28 6mm length 50
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-02900-29
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Dialysis
>
Dialysis Membranes and Tubing
>
Spectra/Por Dialysis Regenerated Cellulose (RC) Membranes
>
Spectra/Por 1 through 5 Standard-Grade Regenerated Cellulose (RC) Dialysis Tubing
>
Spectra/Por 3 Regenerated Cellulose (RC) Dialysis Tubing
Share
|
Spectra/Por 3 Membranes; flat width, 45mm; diameter, 28.6mm; length, 50'
EW-02900-29
$286.00 USD/BOX
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Available in a variety of diameters and flat widths to meet your needs
Regenerated cellulose (RC) membranes for the gentle separation and purification of solutes
MWCO 3500 daltonsDialysis tubing is available in a range of nominal MWCO's for less specific separations. Use the economical-grade Spectra/Por 4 membrane for less critical general applications such as desalting and buffer exchange. All are packaged dry with 10% glycerol and contain trace levels of heavy metals and sulfur.
S/P 1 has MWCO of 6000 to 8000 Dalton
S/P 2 has MWCO of 12,000 to 14,000 Dalton
S/P 3 has MWCO of 3500 Dalton
S/P 4 has MWCO of 12,000 to 14,000 Dalton
s/P 5 has MWCO of 12,000 to 14,000 Dalton
Specifications
Product Type
Dialysis Tubing
Material
Regenerated Cellulose
Membrane MWCO (Daltons)
3500
MWCO (Dalton)
3,500
Flat width (mm)
45
Diameter (mm)
28.6
Length per pack (ft)
50
Brand
Spectra/Por
Manufacturer number
132724
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
High-Performance Dialysis
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title