Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Spectra/Por 4 Membranes; flat width, 10mm; diameter, 6.4mm; length, 100'

EW-02900-41
$360.00 USD/BOX

Usually ships in 12 days.
Qty

  • Available in a variety of diameters and flat widths to meet your application needs
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
MWCO from 12,000 to 14,000 daltons. Similar to SPECTRA/POR 2 membranes, except thicker. Dialysis time is slower, but at much less cost.Dialysis tubing is available in a range of nominal MWCO's for less specific separations. Use the economical-grade Spectra/Por 4 membrane for less critical general applications such as desalting and buffer exchange. All are packaged dry with 10% glycerol and contain trace levels of heavy metals and sulfur.

S/P 1 has MWCO of 6000 to 8000 Dalton
S/P 2 has MWCO of 12,000 to 14,000 Dalton
S/P 3 has MWCO of 3500 Dalton
S/P 4 has MWCO of 12,000 to 14,000 Dalton
s/P 5 has MWCO of 12,000 to 14,000 Dalton
Product Type Dialysis Tubing
Material Regenerated Cellulose
Membrane MWCO (Daltons) 12000
MWCO (Dalton) 12,000 to 14,000
Flat width (mm) 10
Diameter (mm) 6.4
Length per pack (ft) 100
Brand Spectra/Por
Manufacturer number 132697





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title