Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Spectra/Por 7 Membranes; MWCO, 1000; flat width, 45mm; diameter, 28.6mm

EW-02902-58
$266.00 USD/BOX

  • Available in a variety of specific MWCO's
  • Available in a variety of MWCOs
  • Ideal for separations sensitive to heavy metal traces
  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes
For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.These tubing grades offer more defined MWCO's for separation requiring more specific retentivity.
Product Type Dialysis Tubing
Material Regenerated Cellulose
Membrane MWCO (Daltons) 1000
Flat width (mm) 45
Diameter (mm) 28.6
Length per pack (ft) 16
Brand Spectra/Por
Manufacturer number 132105





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title