United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
10 >
Representative photo only
Spooled Masterflex platinum cured silicone tubing L S 16 500 ft
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-96403-16
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Pump Tubing
>
Masterflex L/S Pump Tubing for Masterflex L/S Pumps
>
L/S Spooled Pump Tubing for Masterflex L/S Pumps
>
L/S Silicone (Platinum-Cured) Spooled Pump Tubing for Masterflex L/S Pumps
Share
|
Spooled Masterflex platinum-cured silicone tubing, L/S 16, 500 ft.
SI-96403-16
$923.00 USD/PACKAGE
In stock.
Qty
:
Add to Cart
Save to shopping list
Slightly greater clarity
Smooth surface; low protein binding levels
Preferred by pharmaceutical & biotechnology industries
Fewer potential leachables
USP Class VI; FDA 21 CFR 177.2600
Specifications
Suction lift (m H
2
O)
8.8
Product Type
Pump Tubing
Suction lift (ft H
2
O)
29
Hose barb size (in.)
0.125
Max pressure (bar)
1.725
Max pressure (psi)
25
Max vacuum (mm Hg)
660
Max vacuum (" Hg)
26
Min temperature (º C)
-50
Min temperature (º F)
-58
Max temperature (º C)
230
Max temperature (º F)
446
Tubing ID (mm)
3.2
Tubing ID (in.)
0.12
Length per pack (ft)
500
Pump Series
L/S
Tubing formulation
Silicone
Masterflex tubing size (L/S)
16
Brand
Masterflex
Resin certification (FDA)
Yes
Resin certification (USP)
Yes
Resin certification (3A)
Yes
Max pressure continuous (bar)
2.7
Max pressure continuous (psi)
40
Sterilization Method
Gamma Irradiation
Flow range with 6 to 600 rpm drive (ml/min)
4.8 to 480
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex L/S Tubing Options
Masterflex LS Tubing Life Technical Data
Masterflex® L/S® Precision Pump Tubing
Show All...
Masterflex® L/S® Tubing Life Characteristics
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Peristaltic PumpsMatching the Tubing to the Fluid
Increasing Tubing Life
Masterflex Tubing Life Guidelines
Masterflex Tubing Formulations
Masterflex Connection Newsletter Vol.4-1 - Norprene Food Pump Tubing
Chem-Durance Chemical pump tubing
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title