United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
8 >
Representative photo only
StableTemp Ceramic Stirring Hot Plate 7 x 7 120 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-03407-10
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-27
Innocal NIST Calibration for Stirrers and Heat
$160.00 USD/EACH/EACH
Usually ships in 16 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Stirrers
>
Stirring Hot Plates
>
StableTemp Stirring Hot Plates
Share
|
StableTemp Ceramic Stirring Hot Plate, 7" x 7", 120 VAC
EW-03407-10
$415.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Microprocessor allows for precise control of speed and temperature throughout entire range
Hot Surface Warning safety symbol illuminates when top plate is hot and remains lit until top plate cools below 50°C
Cast-Aluminum base has molded contours to divert liquid spills away from internal components
Instant ON feature starts stirring at 50 RPM to prevent splashing
Units are designed for general laboratory applications, these low profile stirring hotplates feature updated safety features, digital heat display and a two-piece spill resistant cast housing to deflect spills away from critical components. The easy to read digital display shows set point temperature for improved accuracy and repeatability. Built in support rod mount with thumbscrew will accomidate rods up to 13 mm in diameter. Choose corrosion resistant ceramic for high temperature applications or rugged aluminum for temperature uniformity.
What's Included:
detachable 6-ft (1.8-m) cord with plug and PTFE stir bar.
Specifications
Product Type
Stirrer (Hot Plates)
Max load capacity (lbs)
25
Speed (rpm) (low)
30
Speed (rpm) (high)
1200
Max temperature (º C)
540
Top plate material
Ceramic
Top plate dimensions (in.)
7 in x 7 in
Dimensions
8.5 in x 3.75 in x 12.75 in
Max stirring volume (Liters)
4
No. of stirring positions
1
Plug type
U.S. std
Brand
StableTemp
Manufacturer number
1110049SH-CP
Power
120 VAC, 50/60 HZ
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Stirrers and Heat
EW-17002-27
at $160.00 USD/EACH
Usually ships in 16 days.
Add to Cart
Technical Resources
03405-series.pdf
2265_StableTempHotPlatesStirrers_Brochure.pdf
Virtual Catalog (PDFs)
13-14 Catalog
More info
2265.1ZH_StableTempHotPlatesStirrers_Brochure_Chinese.pdf
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title