United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
StableTemp Digital Ceramic Stirring Hot Plate 7 x 7 230 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-03407-45
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-27
Innocal NIST Calibration for Stirrers and Heat
$160.00 USD/EACH/EACH
Usually ships in 16 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Stirrers
>
Stirring Hot Plates
>
StableTemp Digital Stirring Hot Plates
Share
|
StableTemp Digital Ceramic Stirring Hot Plate, 7" x 7", 230 VAC
EW-03407-45
$915.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Microprocessor allows for precise control of speed and temperature throughout entire range
Hot Surface Warning safety symbol illuminates when top plate is hot and remains lit until top plate cools below 50°C
Cast-Aluminum base has molded contours to divert liquid spills away from internal components
Instant ON feature starts stirring at 50 RPM to prevent splashing
These stirring hotplates are designed for applications that require precise accuracy and repeatability. Units feature an updated safety system including a hot surface warning system and over temperature protection. Separate heat and stir displays show actual and set point temperature and stir speed allowing repeatable accurate results. Units include a temperature probe kit which contains a 6” PTFE coated stainless steel probe, 18” support rod, and probe clamp. Timer will shut off heating, stirring or both and sound an audible alarm to alert user.
Over temperature protection system cuts off power to unit in overheat condition protecting both user and sample.
What's Included:
detachable 6-ft cord with plug and PTFE stir bart, temperature probe kit.
Specifications
Product Type
Stirrer (Hot Plates)
Max load capacity (lbs)
25
Speed (rpm) (low)
30
Speed (rpm) (high)
1200
Max temperature (º C)
540
Top plate material
Ceramic
Top plate dimensions (in.)
7 in x 7 in
Dimensions
8.5 in x 3.75 in x 12.75 in
Timer
Yes
Max stirring volume (Liters)
4
No. of stirring positions
1
Plug type
European
CE Compliance
Yes
Brand
StableTemp
Manufacturer number
1130249SHP-CP
Power
240 VAC, 50/60 HZ
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Stirrers and Heat
EW-17002-27
at $160.00 USD/EACH
Usually ships in 16 days.
Add to Cart
Technical Resources
03407-40,-45.pdf
2265.1ZH_StableTempHotPlatesStirrers_Brochure_Chinese.pdf
Virtual Catalog (PDFs)
13-14 Catalog
More info
2265_StableTempHotPlatesStirrers_Brochure.pdf
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title