United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
EW-05053-20
StableTemp
®
vacuum oven, 2.3 cu ft, 120 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-05053-20
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-30
Innocal NIST Calibration for Ovens
$160.00 USD/EACH/EACH
Usually ships in 17 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Ovens
>
Vacuum Ovens
>
StableTemp Vacuum Ovens
Share
|
StableTemp vacuum oven, 2.3 cu ft, 120 VAC
EW-05053-20
$5,040.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Operate at atmospheric pressure or at a vacuum up to 30" Hg
Polycarbonate shield protects door glass from shattering
Ideal for carefully controlled processes such as desiccating, vacuum embedding, plating, and semiconductor processing. Control vacuum levels between 0 and 30" Hg. Silicone door gasket and positive door latch assure a tight seal at all vacuum levels. A three-way valve allows easy evacuation, purging, and venting. The front-mounted stainless steel vacuum line fittings accept 1/4" ID tubing.
Radiant warm-wall heat provides excellent temperature uniformity of ±4°C at 150°C. Use the dial thermometer to monitor the chamber temperature. Insert other types of probes such as gas and humidity into the two rear 1/4" NPT(F) ports to monitor the internal atmosphere. Built-in overtemperature thermostat protects your samples from overheating. Aluminum shelves provide minimal heat retention, reducing hot spots.
What's Included:
a dial thermometer, one-piece shelf assembly with two aluminum shelves, and a 6-ft (1.8-m) three-wire power cord (120 VAC models include plug).
Specifications
Product Type
Vacuum Oven
Capacity (Liters)
56.62
Capacity (cu ft)
2
Chamber size (" W)
14
Chamber size (" H)
14
Chamber size (" D)
20
Minimum temperature (º C)
35
Maximum temperature (º C)
220
Minimum temperature (º F)
95
Maximum temperature (º F)
428
Power (VAC)
120
Temp uniformity
±7.0°C at 100°C
Vacuum display
dial gauge
Display
dial gauge
Vacuum range
0 to 30" Hg
Control type
Hydraulic
Power (Hz)
50/60
Power (Amps)
13.3
Power (watts)
1800
No. of shelf positions
2 (fixed)
No. of shelves
2 (removable)
Brand
Cole-Parmer StableTemp
Construction
Stainless steel interior
Overall dimensions
25"W x 22"H x 25"D
Manufacturer number
3618C/P
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Ovens
EW-17002-30
at $160.00 USD/EACH
Usually ships in 17 days.
Add to Cart
Technical Resources
Cole-Parmer
StableTemp
Ovens and Furnaces Brochure (579 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title