United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
11 >
EW-08021-34
Image is representative of item.
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-08021-34
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Clamps, Supports, Laboratory Jacks
>
Troemner Clamps and Supports
>
Utility Clamps
>
Three-Prong Extension Utility Clamps
Share
|
Standard 3-Prong extension clamp, Dual, Medium, Nickel-plated zinc
EW-08021-34
$41.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Securely hold your flasks, beakers and burettes
An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Designed to securely hold a large variety of glassware and apparatus. Ideal for irregularly shaped objects. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Available in 3 sizes, small, medium, and large. Dual adjustable clamps are offered in either stainless steel or nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers.
Specifications
Product Type
Utility Clamps
Description
Multi-purpose 3 prong single adjust extension clamp
Dimensions (" L)
9.03125
Dimensions (" Dia)
0.43
Opens to dia
2.72 in (69 mm)
Size
Medium
Material (Clamp)
Nickel-plated Zinc
Brand
Troemner
Accessories
Qty
/
Price
Availability
Replacement Sleeves, Red Vinyl, Medium, 3
EW-08027-86
at
$9.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Replacement Sleeves, Black Fiberglass, Medium, 3
EW-08027-96
at
$9.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Thumbscrew Knob
EW-0804795
at
$38.00 USD/EACH
Usually ships in 11 days.
Add to Cart
Replacement Sleeves, Black Vinyl, Medium, 3/pk
EW-08126-34
at
$7.50 USD/PACKAGE
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title