United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-25714-04
Sterile Electroporation Cuvette, 4 mm gap width, green cap, 50/pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-25714-04
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Electroporation
>
Cuvettes for Electroporation
>
Electroporation Cuvettes, Sterile
Share
|
Sterile Electroporation Cuvette, 4 mm gap width, green cap, 50/pack
EW-25714-04
$184.00 USD/PACKAGE
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
High-quality disposable cuvettes are thoroughly cleansed to eliminate trace inhibitor materials
Each cuvette is sterile and individually packed with a presterilized transfer pipette. Color coded frosted caps enable trouble-free labeling and recognition. Sturdy polycarbonate construction withstands pulses of very high voltages or field strength and is autoclavable. Compatible with most electroporation systems.These electroporation cuvettes have a 4mm gap width which allows for low field strength- ideal for mammalian cells.
What's Included:
presterilized transfer pipette.
Specifications
Product Type
Electroporation Cuvettes
Color
Green cap
Application
Lowest field strength for mammilian cells
Gap width
4 mm
Qty/pk
50
Technical Resources
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Isolating Genomic DNA from Whole Blood
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Show All...
Spectrophotometer Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title