Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Sterilized Sampling Bottle Assembly, 500 mL

EW-95644-04
$61.00 USD/EACH

  • Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to process
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
  • Manufactured in an FDA-registered facility from pharmaceutical-grade resins and tubing
Cap assembly includes a seamless liner with premolded C-FLEX® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain assembly sterility until use. Vent tube includes a 0.2-micron Pall® Acrodisc PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.

C-FLEX Tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275°F (–73 to 135°C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
What's Included:
Thermo Scientific Nalgene clear PETG bottle, 38-mm cap, C-FLEX tube/liner assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron vent filter.
Product Type Sterilized Sampling Bottle Assemblies
Volume 500 mL





Technical Resources
Virtual Catalog (PDFs)
  • 11-12 Catalog
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title