log in or register
Stir-Pak High-speed, low-torque mixer system; 23-2300rpm, 115 VAC
EW-50007-32
$1,365.00 USD/EACH
  • Modular system for maximum versatility
  • Modular system for maximum versatility
  • Easily customize your mixer to suit your requirements
  • Easily customize your mixer to suit your requirements
Our modular Stir-Pak® heavy-duty mixer system allows you the flexibility to create the best system for your mixing applications. System components are sold separately, and as convenient, complete system packages—customize your own system or select one of ours.
To customize your own system, simply start with a solid-state speed controller, add one of the four mixing heads depending on your speed and torque requirements, select a shaft and propeller, then choose from clamps and stands that best suit your application. For our full line of laboratory mixing accessories, click here.Our modular Stir-Pak® heavy-duty mixers allow you the flexibility to create the best system for your mixing applications. System components are sold separately, and as convenient, complete system packages—customize your own system or select one of ours.
Product Type Mixer
Brand Stir-Pak
Calibrations
Qty/Price
Availability
at $115.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title