United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Stir Pak speed controller 115 VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-50007-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-03
Innocal NIST Calibration for Mixer
$115.00 USD/EACH/EACH
Usually ships in 13 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Laboratory Mixers
>
Analog Control Laboratory Mixers
>
Stir-Pak Mixers
>
Stir-Pak Heavy-Duty Mixer System
>
Stir-Pak Heavy-Duty Mixer System Controllers
Share
|
Stir-Pak speed controller, 115 VAC
EW-50007-00
$352.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
To customize your own system, simply start with a solid-state speed controller, add one of the
mixing heads
depending on your speed and torque requirements, select a
shaft
and
propeller
, then choose from a
clamp
and
stand
that best suit your application. For our full line of laboratory mixing accessories,
click here
.
Controllers:
Solid-state controller offers precise speed control with a 100:1 adjustment range. A separate ''forward/off/reverse'' switch works independently from the speed control dialallowing you to repeat mixer speed settings from one run to the next. Electronic control maintains speed regulation better than ±2%. Torque limiting provides greater system protection and reduced motor gear failure. An LED indicates when power is on.
Remote controller is connected to the mixer head via a 6-ft (1.8-m), three-wire cordprotects electronics from hazardous spills or splashes. Controller measures 7'' W x 3 3/16'' H x 4 7/8'' D (178 mm W x 81 mm H x 124 mm D).
What's included:
115 VAC model has a 6-ft (1.8-m) cord with grounded plug. The 230 VAC model has an 8-ft (2.4-m) cord with modular line cord and IEC320/CEE22 connector. When ordering, specify country of destination to receive correct line cord.
Specifications
Brand
Stir-Pak
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Mixer
EW-17050-03
at $115.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Technical Resources
Instruction Manual (325 KB)
Cole-Parmer
Stir-Pak
and ServoDyne
Mixers brochure.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title