United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-08758-02
Stopwatch, 1-button, 30 Minute
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-08758-02
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17060-00
NIST-Traceable Calibration; Timer/Stopwatch
$55.00 USD/EACH/EACH
Usually ships in 5 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Timers
>
Stopwatches
>
Mechanical Stopwatches
>
Mechanical Stopwatches
Share
|
Stopwatch, 1-button, 30 Minute
EW-08758-02
$130.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Precision movements are antimagnetic and shock resistant
Stopwatches feature shock absorbers on the balance wheel, temperature-compensated hairspring, and unbreakable mainsprings. Easy-to-read numbers are clearly visible on the large, 1-3⁄4"-diameter face. All watches have locked returns to prevent accidental reset to zero.One-Button stopwatches feature fifteen jewel movement and a single button that starts, stops, and resets the watch. Shatterproof Plexiglas
®
lens offers greater scratch and chemical resistance.
Specifications
Timing capacity
30 min
Product Type
Stopwatches
Case
Nickel-chrome plating
Lens
Shatterproof Plexiglass
Resolution
1/5 sec w/1/100 minute divisions
Movement
Lever
Sweep
60 sec
Dimensions
1.75 in W x 3.5 in D (4.4 x 8.9 cm)
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Timer/Stopwatch
EW-17060-00
at $55.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Basics of Cartridge Filtration
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title