United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
EW-03773-57
LD 25 Dewar pictured with Liquid Withdrawal Device 03773-67 and Roller Base 03773-65 (order each separately)
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-03773-57
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Cryogenics
>
Cryogenic Storage Dewars
>
Taylor Wharton Cryogenic Storage Dewars
Share
|
Storage Dewars, 25 Liters
EW-03773-57
$839.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Superior vacuum performance and insulation provide maximum holding times
Sealed vacuum space between inner and outer shells
Outer shell is constructed of durable, lightweight aluminum
Vacuum space is filled with technologically advanced insulation. Necktube core minimizes liquid nitrogen losses.
Optional roller base lets you easily move larger capacity storage Dewars. Sturdy cast-aluminum frame has five wheels with ball-bearing swivel casters.
Specifications
Product Type
Dewars
Volume (Liters)
25
Dimensions (" H)
25.8
Neck ID
2.5
Static holding time
109 days
Evaporation
0.23 L/day
Brand
Taylor-Wharton
Accessories
Qty
/
Price
Availability
Stainless steel cryogenic transfer hose, 4-ft long
EW-03773-40
at
$215.00 USD/EACH
In stock.
Add to Cart
Stainless steel cryogenic transfer hose, 6-ft long
EW-03773-42
at
$260.00 USD/EACH
In stock.
Add to Cart
Dipper
EW-03773-63
at
$86.00 USD/EACH
In stock.
Add to Cart
Roller base for Dewars with 15-5/8 in. dia base
EW-03773-65
at
$223.00 USD/EACH
In stock.
Add to Cart
Liquid Nitrogen withdrawal system
EW-03773-67
at
$778.00 USD/EACH
In stock.
Add to Cart
Taylor Wharton Storage Dewars Tipping Stand
EW-03773-70
at
$910.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Isolating Genomic DNA from Whole Blood
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title