Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Storage Dewars, 25 Liters

SI-03773-57
$839.00 USD/EACH

  • Superior vacuum performance and insulation provide maximum holding times
  • Sealed vacuum space between inner and outer shells
  • Outer shell is constructed of durable, lightweight aluminum
Vacuum space is filled with technologically advanced insulation. Necktube core minimizes liquid nitrogen losses.

Optional roller base lets you easily move larger capacity storage Dewars. Sturdy cast-aluminum frame has five wheels with ball-bearing swivel casters.
Product Type Dewars
Volume (Liters) 25
Dimensions (" H) 25.8
Neck ID 2.5
Static holding time 109 days
Evaporation 0.23 L/day
Brand Taylor-Wharton

Accessories
Qty/Price
Availability

at $215.00 USD/EACH

In stock.

at $260.00 USD/EACH

In stock.
Dipper
EW-03773-63

at $86.00 USD/EACH

In stock.

at $223.00 USD/EACH

In stock.

at $778.00 USD/EACH

In stock.

at $910.00 USD/EACH

In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title