Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Straight male pipe Adapter, White PP,1/4" OD x 1/8" NPT, 1", 5/8", 7/32"

Item# EW-30815-22

$67.00 USD/PACKAGE of 100

Usually ships in 14 days.
Qty
Product Type Compression Fitting
Material Polypropylene
NPT(M) (in.) 0.125
Tube OD (in.) 0.25
Dimensions orifice 7/32"
Sterilization Ethylene Oxide (ETO)
Dimensions A 1"
Dimensions B 5/8"
Brand Cole-Parmer
Qty/pk 100





Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews