Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Sulfuric Acid, 0.100 Normal (N/10) (20 L)

Ricca Chemical Company  |   Item# WU-86994-56

$137.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Available for order
or call to order
Qty

  • SULFURIC ACID, 0.100 N


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Inorganic Chemicals
Purity grade Components all ACS grade
Normality 0.100
CAS Number Includes 7664-93-9 and 7732-18-5
Form Liquid
Packaging Cubitainer
Synonyms Hygrogen sulfate; Oil of vitriol; Vitriol brown oil; Mattling acid; Battery acid; Sulphuric acid; Electrolyte acid; Dihydrogen sulfate; Spirt of sulphur; Chamber acid
Quantity (mL) 20000
DOT description Not regulated by DOT
Shelf life 24 months
Brand Ricca Chemical Company
Qty/ea 1
Class Titrants





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews