Optional stand has black enameled cast-iron base and zinc-plated rod. Base measures 9"L x 5-1/2"W (229 x 140 mm). Rod measures 24"H (609 mm) x 1/2"dia (13 mm).
Order converter clamp 04714-90 for use with 130-watt ultrasonic processors.
Order light-duty converter clamp 04714-91 for use with 500- and 750-watt ultrasonic processors.
For industrial applications with 500- and 750-watt ultrasonic processors, order heavy-duty converter clamp 04710-94.
Order all converter clamps separately at right.