Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Surface-Cleanse/930 Liquid Concentrate, 1L

EW-18103-00
$33.00 USD/EACH

  • Leaves no charged residue—perfect for cleaning electrical components!
Safely wash away grease, petroleum, fingerprints, oils, scale, oxides, and metallic residues from glass, aluminum, nickel, lead, zinc, ceramic, and plastic without surface damages typically caused by acidic or alkaline cleaners. Use to clean filter membranes and other delicate items.
Non-flammable and non-hazardous under U.S. federal regulations for shipping, storage, and disposal. Meets USDA guidelines for incidental contact with food processing systems. Contains no CFCs, phosphates, silicates, borates, halogens, or phenols.Surface-Cleanse/930 is a gentle, non-ionic cleaner designed to deliver powerful cleaning action. It contains a mixture of nonionic sufacatants that is safe for use on aluminum, zinc and other delicate metals and the cleaner's low sodium content makes it ideal for use on electronic components. Its neutral pH eliminates surface damage that may be caused by an acid or alkaline cleaner.

Target soils include adhesive, oil, flux and ink. Solution is safe for workers and the environment - contains no phosphates, CFCs, ODCs, phenols or borates. Designed for use in CIP, ultrasonic and immersion applications. A 1% to 2% solution is effective in most appilcations. Typical pH: 9.5. NSF registered for USDA A1 use.
Product Type Equipment Cleaner
Format Liquid
Volume (Liters) 1
Packaging 1 Liter
Uses Cleans grease, petroleum, oils from glass, aluminum, lead, zinc and plastic without surface damage
Brand Surface- Cleanse/930
Manufacturer number S2001-12





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title