United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
6 >
EW-86436-00
TechniSat
®
, 70 wipes/tub
Click to enlarge
More Product Details
Specifications
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86436-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Cleaning Supplies
>
Cleaning Wipes
>
Texwipe Low Particulate and Pre-Saturated Wipes
>
TEXWIPE Prewetted Cleanroom Wipes
Share
|
Sorry, this item is now discontinued but has a suggested replacement.
You can compare specifications to the suggested replacement EW-8643601
,
browse similar items
or
Contact us for more information.
TechniSat, 70 wipes/tub
EW-86436-00
$32.00 USD/PACKAGE
Discontinued
Qty
:
Save to shopping list
Save time and avoid solvent spillage with convenient presaturated wipe options
TechniSat
®
Wipes
are nonwoven, 55% cellulose/45% polyester blend to combine the absorbent properties of a natural fiber with the strength and durability of a synthetic wiper. These economical wipes are saturated in 70% isopropyl alcohol/30% DI waterideal for general purpose cleaning or in areas where nonvolatile residue (NVR) is a concern.
Note:
Not available for sale to the state of California
This item is considered a hazardous material.
Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Specifications
Product Type
Wipes
Model
TX1041
Brand
TEXWIPE
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title