United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-80008-55
Temperature/Humidity Strip Chart Recorder
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-80008-55
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-26
NIST-Traceable Calibration; %RH/Temperature Recorder
$160.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Data Loggers and Chart Recorders
>
Humidity Data Loggers and Recorders
>
Temperature/Humidity Strip Chart Recorder
Share
|
Temperature/Humidity Strip Chart Recorder
EW-80008-55
$478.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Alarm and relay features provide added versatility
Vertical, horizontal, free standing or wall mountable
Probes are field interchangeable and can be extended
This temperature, humidity and dew point strip chart recorder is perfect for quality control and assurance for your most critical environments. The unit is menu driven and all parameters are shown on the two line alphanumeric LCD display. Two independent pens record information on a twenty foot long, four and three quarter inch wide strip chart. Chart speeds and temperature ranges are user selectable on the menu driven interface. A full function alarm is provided and can be set to sound an audible signal when the temperature and/or humidity have exceeded an upper or lower set limit.
An alarm delay time can be set prior to activation to prevent any unwanted alarms. Low power normally open contacts are incorporated in the design to allow the activation of a remote alarm or phone dialer. In the event of a power loss a forty-eight hour battery backup is featured to provide continuous operation, and selectable functions are retained in the memory avoiding the re-entering of settings. External power may be supplied by any 12 VDC source such as an automotive, marine or other battery. Each temperature/humidity probe is individually calibrated and field interchangeable.
What’s included: Temperature/Humidity probe with 6-ft cable, one roll of U.S. standard charts, one roll of metric charts, one red pen, one blue pen, and battery holder.
Specifications
Product Type
Temperature/Humidity Strip Chart Recorders
Min temperature (º C)
-40
Max temperature (º C)
60
Min temperature (º F)
-40
Max temperature (º F)
150
Dew point accuracy
±2°F (±1°C)
Alarm
Yes
Dew point resolution
1°F (0.5°C)
Dew point range
32 to 125°F (0 to 50°C)
Temp resolution
0.1°F/°C
Chart type
Paper-Strip
Accuracy
Temperature accuracy: ±2°F (±1°C); Humidity accuracy: ±2% from 0 to 95% RH; Dew Point accuracy: ±2°F (±1°C)
Display
16 characters, 2-line LCD
Chart speed
0.25, 0.5, 1, 2 in./min; 0.25, 1, 2, 4, 8 in./hr; 0.5, 2, 4, 8, 16 cm/hr; 0.5, 1, 2, 4 cm/min (user selectable)
CE Compliance
Yes
No. of pens
2
Dimensions
7.25 in W x 9.25 in H x 2 in D (18.4 x 23.5 x 5.1 cm)
Power
12 VDC
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
220 VAC power adapter
EW-80008-60
at
$21.75 USD/EACH
Usually ships in 14 days.
Add to Cart
High temperature thermocouple probe, 100-ft.
EW-80008-65
at
$76.00 USD/EACH
Usually ships in 14 days.
Add to Cart
Chart paper, U.S. standard, 4/bx.
EW-80009-75
at
$20.75 USD/BOX
Usually ships in 14 days.
Add to Cart
Chart paper, metric, 4/bx.
EW-80009-80
at
$20.75 USD/BOX
In stock.
Add to Cart
Blue Pen for Humidity measurement, 2/pk
EW-80055-70
at
$20.25 USD/PACKAGE of 2
In stock.
Add to Cart
Red Pen for Temperature measurement, 2/pk
EW-80055-71
at
$20.25 USD/PACKAGE of 2
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; %RH/Temperature Recorder
EW-17030-26
at $160.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
Related technical articles (Roll over for summary)
Tech Challenge: Temperature Monitoring of Food Products
Recorder Selection Guide
Temperature FAQs
Show All...
Temperature Instrument Ranges
Getting a Handle on Temperature
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title