Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Thermo-Dynamic Steam Trap, 3/4"NPT(F), 600 psi

Spirax Sarco  |   Item# EW-07600-06

$545.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.

Usually ships in 7 days.

  • The tight shut-off prevents valuable steam from being wasted
  • Cost effective, compact design makes them ideal for drainage of steam mains, steam tracing lines, and small process equipment where size and efficient operation are important.
  • Discharge from Thermo-Dynamic (TD) traps is close to steam temperature — steam space is kept free from condensate.
  • Constructed of stainless steel, TD traps withstand superheat, waterhammer, freezing, corrosive condensate, and vibration.
Note: Minimum pressure for satisfactory operation is 3.5 psig. Maximum back pressure should not exceed 80% of the inlet pressure under any conditions of operation, otherwise the trap may not shut.Typical applications:
  • Steam main drain and tracer lines
  • Process equipment
  • Laundry and kitchen equipment
  • Superheated steam applications
  • Outdoor installations subject to freezing
Product Type Steam handling fittings
Material Stainless Steel
Size 3/4" NPT(F)
NPT(F) 0.75
Max operating pressure 600 psig
Max temperature 800°F (427°C)
Brand Spirax Sarco
Qty/ea 1
Model TD52 3/4" 54531C

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews