| Product Type |
Metal Detectors |
| Operating conditions |
14 to 104°F (-10 to 40°C), 20 to 80% RH noncondensing |
| Power |
85 to 260 VAC single phase plus earth ground 47 to 65 Hz, 100 watts max |
| Outputs |
Six relays: 250 VAC, 2 A max; 50 VDC, 1 A max |
| Output allocation |
(Selectable via menu) Reject 1, reject 2, fault, alarm, warning, QA lamp |
| Inputs |
Six: active 12 VDC + 12 V auxiliary supply for input sensors |
| Input allocation |
(Selectable via menu) Speed sensor, keylock, product select 1, product select 2, infeed PEC, reject confirmation 1 (bin full), reject confirmation 2, external suppression |
| Construction |
304 stainless steel straightlined case, ABS plastic control panel |
| Protection ratings |
IP69k, NEMA 4, ATEX zone 22, hazardous location Class II, Division 2, Groups F & G |
| Languages |
English, Spanish, French, German, Italian, Dutch, Chinese, Russian, Czech, Polish |
| Aperture height |
125 mm |
| Aperture width |
200 mm |
| Product speed |
1.7 to 3300 ft/min (0.5 to 1000 m/min) |
| CE Compliance |
Yes |
| Manufacturer number |
APEX 100, 200 x 125 mm |
| Brand |
Thermo Scientific |