Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Thermo Scientific APEX 100 Metal Detector, with 500 mm width x 200 mm height aperture

EW-20039-68
$10,803.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Ensure food safety by detecting all types of metal contaminants
  • Proprietary multicoil design provides excellent sensitivity to all metals
  • Fully tested to meet IP69k/NEMA/ATEX requirements
  • Application flexibility with dual frequency and gain operation
  • Remove product effects with traditional phasing or unique Intellitrack XR (IXR)
  • Quickly set up products with simple autolearn function
  • Multilingual help text
  • Multi-level password protection to ensure correct operation in production
  • Fe, non-Fe, and 316 stainless steel test cards included
  • Rated for IP69k washdown and dust environments
The APEX 100 metal detector provides basic, cost-effective protection from the most common metal contaminants found in food production today. This metal detector is instrumental in your HACCP policy compliance and is designed to perform for years with minimal maintenance.

To deliver greater accuracy in detecting contaminants in wet and salty products, the APEX 100 features the innovative Intellitrack XR (IXR) technology. This technology learns and cancels the sinusoidal X (reactive) and R (resistive) product signals, and only detects the metal contaminants. To maintain high accuracy over time, the IXR tracks product changes and adjusts the product signals to eliminate the risk of contaminants going undetected.

Installation and setup is quick and easy, and no special training is required. The detectors come with everything you need, including electrical fittings as well as clear wiring and mechanical mounting instructions. An easy-to-use icon-driven interface features a step-by-step quick-reference guide, multilingual help text, and autocalibration.

Optimize performance by matching the metal detector exactly to your conveyor width and product height with the numerous aperture size configurations available. Easily customize your metal detector with additional accessories such as MODBUS communications modules, field compression flanges, remote control mounting kit, alarm/fault warning lights and horn, and photo eyes for product inspection trigger.

Note: To ensure proper aperture clearance, add at least 1.9 in (50 mm), to the width and height of the product. The control panel is always mounted in the center of the front of the metal detector case unless remote mounting is ordered.
What's Included:
metal detector head, mounting hardware, watertight electrical fittings, test cards, and documentation on CD (users' guides and installation instructions, quick reference guide, and PC software simulator)
Product Type Metal Detectors
Operating conditions 14 to 104°F (-10 to 40°C), 20 to 80% RH noncondensing
Power 85 to 260 VAC single phase plus earth ground 47 to 65 Hz, 100 watts max
Outputs Six relays: 250 VAC, 2 A max; 50 VDC, 1 A max
Output allocation (Selectable via menu) Reject 1, reject 2, fault, alarm, warning, QA lamp
Inputs Six: active 12 VDC + 12 V auxiliary supply for input sensors
Input allocation (Selectable via menu) Speed sensor, keylock, product select 1, product select 2, infeed PEC, reject confirmation 1 (bin full), reject confirmation 2, external suppression
Construction 304 stainless steel straightlined case, ABS plastic control panel
Protection ratings IP69k, NEMA 4, ATEX zone 22, hazardous location Class II, Division 2, Groups F & G
Languages English, Spanish, French, German, Italian, Dutch, Chinese, Russian, Czech, Polish
Aperture height 200 mm
Aperture width 500 mm
Product speed 1.7 to 3300 ft/min (0.5 to 1000 m/min)
CE Compliance Yes
Manufacturer number APEX 100, 500 x 200 mm
Brand Thermo Scientific





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title