United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-99564-10
THERMO AQUAMATE 7000 VIS SPECTROMETER
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Technical information
Instruction manual
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-99564-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Benchtop Spectrometers
>
Thermo Scientific AquaMate 7000 Vis and 8000 UV-Vis Spectrophotometers
Share
|
Thermo Scientific AquaMate 7000 Vis Spectrometer
EW-99564-10
$3,750.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Designed for water and wastewater analysis
Over 260 pre-programmed water quality methods plus user customizable methods
Methods stored on USB stick to be added or removed as needed
One-point calibration adjustment ensures the highest accuracy
Compatible with several leading brands of reagents
AquaMate spectrophotometers offer the accuracy, dependability and ease of use your lab requires. Compact design with intuitive operation targeted to water and wastewater analysis. The pre-programmed or user-customizable water quality methods offer absorbance, % transmittance and concentration testing. Ensure wavelength accuracy and quality measurements with performance verification testing in accordance to GLP and GMP.
Aquamate features three USB connectors, providing the option to store methods and data to a USB stick, transfer data to a computer, or print data to an external printer. A wide range of vial sizes can be used with the 3 position cell-holder (turret): 13 to 25 mm round vials, 20 to 50 mm rectangular vials, and a 10 mm square vial. System is compatible with Orion, Merck, Chemetrics, and Hach chemistries.
AQ7000 Vis Spectrophotometer
is the cost-effectve solution for accurate visible measurements. It offers 325 to 1100 nm selectable wavelengths, ideal for most concentration measurements. Combines an intuitive user interface, tungsten-halogen lamp, and simple single-beam optical geometry for accurate, reproducible results.
What's Included:
methods on USB stick, three-position cell holder (turret), USB cable, dust cover, and 110V and 220V power cords.
AQ8000 adds: six-position and single-position cell holder (turret).
Specifications
Product Type
Visible Spectrophotmeter
Bandwidth
5 nm
Wavelength range
325 - 1100 nm
Wavelength accuracy
+/- 1 nm
Photometric range
0 - 125%; 0 to 3 A; +/- 9999 C
Photometric drift
<0.1% T at 340 and 400 nm
Detector
Silicon photdiode
Output
USB
Beam Type
Single
Applications
Water and Wastewater
Stray light
0.002 A/hr after warm-up
Source lamp
Tungsten-halogen
Lamp life
1000 hrs
Display
Graphic, backlit LCD
Dimensions (in.)
11 3/4 x 9 2/3 x 15 3/4
Dimensions (cm)
30 x 25 x 40
Model
7000
Power (VAC)
100 to 240
Power (Hz)
50/60
Brand
Thermo Scientific
Accessories
Qty
/
Price
Availability
AquaMate 6-Position Cell Holder
EW-99564-56
at
$215.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate 3-Position Cell Holder
EW-99564-58
at
$450.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate 1-Position Cell Holder
EW-99564-60
at
$140.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate USB Memory Stick
EW-99564-61
at
$45.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate Power Cord, 240V, UK Plug
EW-99564-62
at
$30.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate Power Cord, 220V, Euro Plug
EW-99564-63
at
$30.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate Power Cord, 110V, US Plug
EW-99564-64
at
$30.00 USD/EACH
Usually ships in 8 days.
Add to Cart
AquaMate Power Cord, China Plug
EW-99564-65
at
$30.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
AquaMate User Guide
AquaMate Method List
AQUAfast Reagent Instructions for AquaMate
More info
AquaMate and Star A Promotion
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title