Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Thermo Scientific Barnstead Half-Sized Holder System
Thermo Scientific Barnstead  |   Mfr# D5839  |   Item# EW-99259-00
$497.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
  • Modular design - fill with your choice of cartridges
  • Perfect for general pretreatment of laboratory water
B-PURE™ systems are modular wall-mountable systems for point-of-use deionization. Simply add modules as needed when your water requirements change. All systems accept pre- and posttreatment cartridges. Use pretreatment cartridges to filter water before reverse osmosis, deionization, or distillation. Use posttreatment cartridges at the point of use for filtration of ultrapure water. Easily install and remove cartridges; a quarter twist releases canister. A PURA®-Lite resistivity indicator can be mounted directly to the housing to monitor water purity threshold—order separately.
Thermo Scientific Barnstead Half-Sized Holder System is a compact stand-alone unit or a space-saving addition to your existing B-PURE system. System accepts the pre- and posttreatment filter cartridges sold separately. Canister measures 10" in length.
Product Type Half-sized
Max flow rate 4 liters per minute
Inlet pressure (min) 5 psi
Inlet pressure (Max) 100 psi
Inlet port 1/2" NPT(F)
Inlet water temperature 40 to 120F (4 to 49C)
Dimensions 7"W x 14"H x 6-5/8"D
Model D5839
Qty/ea 1
Brand Thermo Scientific Barnstead
Manufacturer number D5839
Accessories
Qty/Price
Availability
at $97.00 USD/EACH
Usually ships in 12 days.
at $425.00 USD/EACH
Usually ships in 12 days.
at $419.00 USD/EACH
Usually ships in 12 days.
at $97.00 USD/EACH
Usually ships in 13 days.
at $97.00 USD/EACH
Usually ships in 13 days.
at $97.00 USD/EACH
Usually ships in 13 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456