Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register
Thermo Scientific Barnstead Pretreatment Cartridge for B-Pure, E-Pure, Nanopure
Thermo Scientific Barnstead  |   Mfr# D0835  |   Item# EW-01505-22
$175.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Product Type Water Purification Cartridge
Capacity As determined by feed water quality
Compatible with All NANOpure™ E-pure™, B-pure™ systems (except Nanopure Infinity)
Capacity (Liters) 18,925
Function Removes colloids, chlorine, and organics; use when nonionized solids and/or colloids are present
Capacity (gal.) 5000
Qty/ea 1
Brand Thermo Scientific Barnstead
Manufacturer number D0835
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456